Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
101107268
Gene Name
interleukin 26
Gene Aliases
-
Chromosome Location
Chr.3: 162626709 - 162644091 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006486.4 XP_004006535.1 2 - 3 247 118

View Full Product Details