Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GZMK
Entrez Gene ID
101117363
Gene Name
granzyme K
Gene Aliases
-
Chromosome Location
Chr.16: 25450250 - 25460559 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016992.3 XP_004017041.2 4 - 5 635 63

View Full Product Details