Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSN2
Entrez Gene ID
443391
Gene Name
casein beta
Gene Aliases
-
Chromosome Location
Chr.6: 94727455 - 94736605 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009373.1 NP_001009373.1 6 - 7 237 74
GenBank mRNA X16482.1 - - 237 74

View Full Product Details