Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK4
Entrez Gene ID
100144756
Gene Name
cyclin dependent kinase 4
Gene Aliases
-
Chromosome Location
Chr.3: 173531274 - 173534349 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127269.1 NP_001120741.1 6 - 7 820 67
XM_012158548.3 XP_012013938.2 7 - 8 1039 67
GenBank mRNA EU626223.1 - 6 - 7 820 67
FJ943997.1 - 6 - 7 820 67

View Full Product Details