Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICAM1
Entrez Gene ID
443045
Gene Name
intercellular adhesion molecule 1
Gene Aliases
ICAM-1
Chromosome Location
Chr.5: 13147267 - 13157757 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009731.1 NP_001009731.1 5 - 6 1197 73
XM_027969188.1 XP_027824989.1 4 - 5 2050 73
GenBank mRNA AF110984.1 - 5 - 6 1197 73

View Full Product Details