Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CETN2
Entrez Gene ID
100302545
Gene Name
centrin 2
Gene Aliases
-
Chromosome Location
Chr.X: 90079173 - 90082345 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163594.2 NP_001157066.1 1 - 1 294 114
GenBank mRNA FJ937949.1 - 1 - 1 294 114

Gene Symbol
LOC114111037
Entrez Gene ID
114111037
Gene Name
centrin-2
Gene Aliases
-
Chromosome Location
Chr.26: 1349257 - 1350285 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962657.1 XP_027818458.1 1 - 1 342 114

View Full Product Details