Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYZL4
Entrez Gene ID
100579170
Gene Name
lysozyme like 4
Gene Aliases
-
Chromosome Location
Chr.19: 15547263 - 15551885 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246007.1 NP_001232936.1 3 - 4 378 121
GenBank mRNA HQ285240.1 - 3 - 4 378 121

View Full Product Details