Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDHHC7
Entrez Gene ID
100302557
Gene Name
zinc finger DHHC-type palmitoyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.14: 11786658 - 11815195 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163604.1 NP_001157076.1 4 - 5 613 103
XM_027977366.1 XP_027833167.1 6 - 7 103
XM_027977367.1 XP_027833168.1 6 - 7 103
GenBank mRNA FJ937952.1 - 4 - 5 613 103

View Full Product Details