The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004002745.4 | XP_004002794.1 | 4 - 5 | 651 | 91 |
XM_012185836.3 | XP_012041226.1 | 4 - 5 | 755 | 91 | |
XM_027977940.1 | XP_027833741.1 | 4 - 5 | 671 | 91 |