Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF3L2
Entrez Gene ID
105607095
Gene Name
outer dense fiber of sperm tails 3 like 2
Gene Aliases
-
Chromosome Location
Chr.5: 45246211 - 45250837 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970616.1 XP_027826417.1 1 - 2 268 104

View Full Product Details