Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP62CL
Entrez Gene ID
101115862
Gene Name
nucleoporin 62 C-terminal like
Gene Aliases
-
Chromosome Location
Chr.X: 136481970 - 136589246 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012107591.3 XP_011962981.1 7 - 8 845 100
XM_012107592.3 XP_011962982.1 7 - 8 848 100

View Full Product Details