Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2E3
Entrez Gene ID
101121069
Gene Name
ubiquitin conjugating enzyme E2 E3
Gene Aliases
-
Chromosome Location
Chr.2: 139068007 - 139162191 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012132197.3 XP_011987587.1 4 - 5 495 122
XM_012132205.3 XP_011987595.1 4 - 5 574 122
XM_027964786.1 XP_027820587.1 4 - 5 757 122

View Full Product Details