Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFS4
Entrez Gene ID
101118899
Gene Name
NADH:ubiquinone oxidoreductase subunit S4
Gene Aliases
-
Chromosome Location
Chr.16: 26758797 - 26872208 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016997.4 XP_004017046.1 4 - 5 450 104

View Full Product Details