Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAZF1
Entrez Gene ID
101113233
Gene Name
JAZF zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.4: 74856456 - 75188631 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095333.2 XP_014950819.2 4 - 5 738 79

View Full Product Details