Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSTC
Entrez Gene ID
101121263
Gene Name
oligosaccharyltransferase complex non-catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.6: 19762186 - 19773513 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009629.4 XP_004009678.1 3 - 4 502 105

View Full Product Details