Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AEN
Entrez Gene ID
101116260
Gene Name
apoptosis enhancing nuclease
Gene Aliases
-
Chromosome Location
Chr.18: 16840299 - 16851967 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957053.1 XP_027812854.1 2 - 3 670 60

View Full Product Details