Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPCEF1
Entrez Gene ID
101107028
Gene Name
interaction protein for cytohesin exchange factors 1
Gene Aliases
-
Chromosome Location
Chr.8: 85364207 - 85487079 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012183166.3 XP_012038556.2 7 - 8 1072 96
XM_027972591.1 XP_027828392.1 9 - 10 982 96
XM_027972592.1 XP_027828393.1 9 - 10 959 96

View Full Product Details