Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CATSPER3
Entrez Gene ID
101104315
Gene Name
cation channel sperm associated 3
Gene Aliases
-
Chromosome Location
Chr.5: 48469580 - 48504199 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970618.1 XP_027826419.1 3 - 4 292 125

View Full Product Details