Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCR3
Entrez Gene ID
101122313
Gene Name
natural cytotoxicity triggering receptor 3
Gene Aliases
-
Chromosome Location
Chr.20: 29509369 - 29542626 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018959.4 XP_004019008.2 4 - 5 1065 65
XM_012100954.3 XP_011956344.1 5 - 6 992 65
XM_027958851.1 XP_027814652.1 5 - 6 896 65
XM_027958852.1 XP_027814653.1 4 - 5 619 65
XM_027958853.1 XP_027814654.1 4 - 5 850 65

View Full Product Details