Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD3
Entrez Gene ID
101115608
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.X: 136096332 - 136172144 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963490.1 XP_027819291.1 6 - 7 659 107

View Full Product Details