Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARGFX
Entrez Gene ID
101118848
Gene Name
arginine-fifty homeobox
Gene Aliases
-
Chromosome Location
Chr.1: 204164348 - 204170689 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962705.1 XP_027818506.1 2 - 3 273 78

View Full Product Details