Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK11
Entrez Gene ID
101113783
Gene Name
kallikrein related peptidase 11
Gene Aliases
-
Chromosome Location
Chr.14: 60499457 - 60504953 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977541.1 XP_027833342.1 5 - 6 1494 72

View Full Product Details