Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTUD6B
Entrez Gene ID
101114028
Gene Name
OTU deubiquitinase 6B
Gene Aliases
-
Chromosome Location
Chr.9: 95010880 - 95028863 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011844.3 XP_004011893.3 3 - 4 421 70

View Full Product Details