Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP298
Entrez Gene ID
101102107
Gene Name
cilia and flagella associated protein 298
Gene Aliases
C1H21orf59
Chromosome Location
Chr.1: 131039204 - 131052107 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002768.3 XP_004002817.2 3 - 4 1119 140
XM_012186326.2 XP_012041716.2 3 - 4 1131 140
XM_027978278.1 XP_027834079.1 3 - 4 310 140

View Full Product Details