Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK16
Entrez Gene ID
101105955
Gene Name
potassium two pore domain channel subfamily K member 16
Gene Aliases
-
Chromosome Location
Chr.20: 14747546 - 14754042 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019206.4 XP_004019255.2 2 - 3 319 82

View Full Product Details