Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DCDC2C
Entrez Gene ID
101114755
Gene Name
doublecortin domain containing 2C
Gene Aliases
-
Chromosome Location
Chr.2: 2057983 - 2109918 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964045.1 XP_027819846.1 8 - 9 951 68

View Full Product Details