Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBLN3
Entrez Gene ID
101112982
Gene Name
cerebellin 3 precursor
Gene Aliases
-
Chromosome Location
Chr.7: 22037697 - 22040970 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010296.4 XP_004010345.1 1 - 2 1050 64

View Full Product Details