Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBXIP1
Entrez Gene ID
101104627
Gene Name
PBX homeobox interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 111325896 - 111342427 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975818.1 XP_027831619.1 2 - 3 363 83
XM_027975824.1 XP_027831625.1 2 - 3 363 83
XM_027975833.1 XP_027831634.1 2 - 3 175 83

View Full Product Details