Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDBF2
Entrez Gene ID
101105062
Gene Name
zinc finger DBF-type containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 222523227 - 222533610 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964846.1 XP_027820647.1 2 - 3 187 84

View Full Product Details