Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHURC1
Entrez Gene ID
105607024
Gene Name
churchill domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 80471780 - 80489533 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012181836.3 XP_012037226.2 2 - 3 254 80

View Full Product Details