Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB1
Entrez Gene ID
101109805
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 80083310 - 80108045 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010718.4 XP_004010767.1 2 - 3 2003 66

View Full Product Details