Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKH
Entrez Gene ID
101103426
Gene Name
ANKH inorganic pyrophosphate transport regulator
Gene Aliases
-
Chromosome Location
Chr.16: 63657483 - 63825756 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027980157.1 XP_027835958.1 3 - 4 713 87

View Full Product Details