Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101106665
Entrez Gene ID
101106665
Gene Name
NUT family member 2G
Gene Aliases
FAM22D
Chromosome Location
Chr.2: 24066356 - 24072505 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964014.1 XP_027819815.1 5 - 6 1365 89

View Full Product Details