Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101115856
Entrez Gene ID
101115856
Gene Name
sperm acrosome-associated protein 5
Gene Aliases
SPACA5
Chromosome Location
Chr.X: 59049221 - 59053067 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022132.4 XP_004022181.1 1 - 2 1750 80

View Full Product Details