Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL27
Entrez Gene ID
101111931
Gene Name
methyltransferase like 27
Gene Aliases
WBSCR27
Chromosome Location
Chr.24: 34186088 - 34190350 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961648.1 XP_027817449.1 5 - 6 517 85

View Full Product Details