Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM92
Entrez Gene ID
101107614
Gene Name
transmembrane protein 92
Gene Aliases
-
Chromosome Location
Chr.11: 26918079 - 26938172 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012772.4 XP_004012821.1 3 - 4 592 61
XM_015098714.2 XP_014954200.1 3 - 4 592 61
XM_027974708.1 XP_027830509.1 3 - 4 592 61
XM_027974709.1 XP_027830510.1 3 - 4 277 61

View Full Product Details