Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNN3
Entrez Gene ID
101112614
Gene Name
potassium calcium-activated channel subfamily N member 3
Gene Aliases
-
Chromosome Location
Chr.1: 111072297 - 111243615 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002559.4 XP_004002608.2 7 - 8 2284 73
XM_027975753.1 XP_027831554.1 7 - 8 2284 73
XM_027975761.1 XP_027831562.1 7 - 8 1668 73
XM_027975769.1 XP_027831570.1 7 - 8 1541 73

View Full Product Details