Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WHRN
Entrez Gene ID
101118591
Gene Name
whirlin
Gene Aliases
-
Chromosome Location
Chr.2: 9586111 - 9676699 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964085.1 XP_027819886.1 4 - 5 1295 84

View Full Product Details