Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PRX
Entrez Gene ID
101119499
Gene Name
periaxin
Gene Aliases
-
Chromosome Location
Chr.14: 52940917 - 52955914 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978312.1 XP_027834113.1 4 - 5 2386 79
XM_027978313.1 XP_027834114.1 5 - 6 487 79

View Full Product Details