Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBR4
Entrez Gene ID
101122167
Gene Name
carbonyl reductase 4
Gene Aliases
-
Chromosome Location
Chr.2: 121266503 - 121286441 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093286.2 XP_014948772.2 1 - 2 323 107

View Full Product Details