Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
101117467
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 30136699 - 30147785 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003543.3 XP_004003592.2 2 - 3 332 112

View Full Product Details