Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SS18
Entrez Gene ID
101117893
Gene Name
SS18 subunit of BAF chromatin remodeling complex
Gene Aliases
-
Chromosome Location
Chr.23: 35454925 - 35527182 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012103457.3 XP_011958847.2 4 - 5 394 109
XM_015103616.2 XP_014959102.2 4 - 5 430 109
XM_015103617.2 XP_014959103.2 4 - 5 430 109

View Full Product Details