Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD45
Entrez Gene ID
101116848
Gene Name
ankyrin repeat domain 45
Gene Aliases
-
Chromosome Location
Chr.12: 59117126 - 59162827 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012187544.3 XP_012042934.1 2 - 3 446 97

View Full Product Details