Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101101992
Entrez Gene ID
101101992
Gene Name
trace amine-associated receptor 9
Gene Aliases
TAAR9
Chromosome Location
Chr.8: 63966039 - 63967085 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011328.3 XP_004011377.2 1 - 1 115 94

View Full Product Details