Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTOP2
Entrez Gene ID
101102159
Gene Name
otopetrin 2
Gene Aliases
-
Chromosome Location
Chr.11: 6780758 - 6790963 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975402.1 XP_027831203.1 5 - 6 1066 70

View Full Product Details