Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS26C
Entrez Gene ID
101116695
Gene Name
VPS26 endosomal protein sorting factor C
Gene Aliases
DSCR3
Chromosome Location
Chr.1: 292711966 - 292738498 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003405.3 XP_004003454.1 7 - 8 884 90
XM_015092705.2 XP_014948191.1 7 - 8 1236 90

View Full Product Details