Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMCR8
Entrez Gene ID
101120176
Gene Name
SMCR8-C9orf72 complex subunit
Gene Aliases
-
Chromosome Location
Chr.11: 28953020 - 28961922 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012734.3 XP_004012783.1 1 - 2 2801 109

View Full Product Details