Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM86A
Entrez Gene ID
101121641
Gene Name
transmembrane protein 86A
Gene Aliases
-
Chromosome Location
Chr.21: 26494621 - 26499017 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012101794.3 XP_011957184.1 1 - 2 2036 69
XM_027959607.1 XP_027815408.1 2 - 3 408 69

View Full Product Details