Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRR1
Entrez Gene ID
101114365
Gene Name
gamma-aminobutyric acid type A receptor subunit rho1
Gene Aliases
-
Chromosome Location
Chr.8: 53860832 - 53879267 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972716.1 XP_027828517.1 3 - 4 324 100

View Full Product Details