Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ART4
Entrez Gene ID
101117410
Gene Name
ADP-ribosyltransferase 4 (inactive) (Dombrock blood group)
Gene Aliases
-
Chromosome Location
Chr.3: 215188754 - 215207684 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012175310.3 XP_012030700.2 1 - 2 340 77

View Full Product Details